Gene Transcripts

Gene Symbol
VIPAS39
Entrez Gene ID
63894
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
Gene Aliases
C14orf133, SPE-39, SPE39, VIPAR, VPS16B, hSPE-39
Chromosome Location
Chr. 14: 77426675 - 77457640 on Build GRCh38
UniGene ID
Hs.16157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193314.1 NP_001180243.1 15, 16 1642
NM_001193315.1 NP_001180244.1 15, 16 1242
NM_001193316.1 NP_001180245.1 14, 15 1095
NM_001193317.1 NP_001180246.1 15, 16 1419
NM_022067.3 NP_071350.2 16, 17 1513
XM_011537066.1 XP_011535368.1 14, 15 1140
XM_017021579.1 XP_016877068.1 14, 15 1274
XM_017021580.1 XP_016877069.1 15, 16 1233
XM_017021581.1 XP_016877070.1 15, 16 1367
GenBank mRNA AK022769.1 - 15, 16 1190
AK022925.1 - 15, 16 1168
AK024661.1 - 4, 5 338
AK098254.1 - 15, 16 1603
AK222706.1 - 15, 16 1188
AK297388.1 - 15, 16 1344
AK298354.1 - 14, 15 1041
BC015054.1 - 16, 17 1434 3920459
CR457294.1 - 14, 15 1096

View Full Product Details