Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195139.1 | NP_001182068.1 | 16 | 1975 | |
NM_015386.2 | NP_056201.2 | 17 | 2038 | ||
XM_011522981.2 | XP_011521283.1 | 18 | 2198 | ||
GenBank mRNA | AB088369.1 | - | 18 | 2174 | |
AK022874.1 | - | 17 | 2018 | ||
AK026435.1 | - | 13 | 1332 | ||
AK096557.1 | - | 15 | 1756 | ||
AK293235.1 | - | 14 | 1582 | ||
AK297557.1 | - | 18 | 1981 | ||
AL050101.1 | - | 13 | 1340 | ||
BC000796.1 | - | 1 | 36 | ||
BC006306.2 | - | 14 | 1554 | ||
BC013347.2 | - | 13 | 1346 | ||
BC027726.2 | - | 16 | 1949 | ||
BC072438.1 | - | 17 | 2018 | 6011802 | |
BX648010.1 | - | 15 | 2706 |