Gene Transcripts

Gene Symbol
TMEM240
Entrez Gene ID
339453
Gene Name
transmembrane protein 240
Gene Aliases
C1orf70, SCA21
Chromosome Location
Chr. 1: 1534778 - 1540360 on Build GRCh38
UniGene ID
Hs.668654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114748.1 NP_001108220.1 4 477
GenBank mRNA BC147018.1 - 5 506
BC147020.1 - 5 506 9056948
BU929336.1 - 4 458

View Full Product Details