Gene Transcripts

Gene Symbol
PURA
Entrez Gene ID
5813
Gene Name
purine rich element binding protein A
Gene Aliases
MRD31, PUR-ALPHA, PUR1, PURALPHA
Chromosome Location
Chr. 5: 140114123 - 140119416 on Build GRCh38
UniGene ID
Hs.443121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005859.4 NP_005850.1 1 1403
GenBank mRNA BC036087.1 - 1 1356
X91648.1 - 1 377

View Full Product Details