Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007123.5 | NP_009054.5 | 17, 18 | 4191 | |
NM_206933.2 | NP_996816.2 | 17, 18 | 4191 | ||
GenBank mRNA | AF055580.2 | - | 17, 18 | 4191 | |
AY481573.1 | - | 17, 18 | 4191 |