Gene Transcripts

Gene Symbol
C4orf3
Entrez Gene ID
401152
Gene Name
chromosome 4 open reading frame 3
Gene Aliases
Chromosome Location
Chr. 4: 119296419 - 119304445 on Build GRCh38
UniGene ID
Hs.173705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001701.3 NP_001001701.2 2 856
NM_001170330.1 NP_001163801.1 3 977
GenBank mRNA AK024537.1 - 1 52
AK025703.1 - 2 620
BC017399.1 - 2 679 4108498
CN361054.1 - 2 407

View Full Product Details