Gene Transcripts
- Gene Symbol
- GORAB
- Entrez Gene ID
- 92344
- Gene Name
- golgin, RAB6 interacting
- Gene Aliases
- GO, NTKLBP1, SCYL1BP1
- Chromosome Location
- Chr. 1: 170532122 - 170553834 on Build GRCh38
- UniGene ID
- Hs.183702
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB256891.1 |
- |
4 |
508 |
|
Chr. 1 - 170532386 - 170544785 [+] |
GenBankmRNA |
DB256893.1 |
- |
4 |
508 |
|
Chr. 1 - 170532386 - 170544781 [+] |
View Full Product Details