Gene Transcripts

Gene Symbol
SAMD12
Entrez Gene ID
401474
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
Chromosome Location
Chr. 8: 118131828 - 118621995 on Build GRCh38
UniGene ID
Hs.359393
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101676.1 NP_001095146.1 3 395
NM_207506.2 NP_997389.2 3 395
NR_109794.1 3 310
XM_011517031.2 XP_011515333.1 3 435
XM_011517034.2 XP_011515336.1 3 434
XM_011517035.2 XP_011515337.1 3 434
XM_017013409.1 XP_016868898.1 3 437
XM_017013410.1 XP_016868899.1 3 435
XM_017013411.1 XP_016868900.1 3 435
XM_017013412.1 XP_016868901.1 3 435
GenBank mRNA AK096777.1 - 3 331
AK311704.1 - 3 310
BC121818.1 - 3 328 40109130
BC124554.1 - 3 328 40109129
DA335826.1 - 3 395
DA459013.1 - 3 374
DB228621.1 - 3 310

View Full Product Details