Gene Transcripts

Gene Symbol
PDXDC2P
Entrez Gene ID
283970
Gene Name
pyridoxal dependent decarboxylase domain containing 2, pseudogene
Gene Aliases
NPIP, PDXDC2
Chromosome Location
Chr. 16: 69976297 - 70065948 on Build GRCh38
UniGene ID
Hs.513695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003610.1 25 2640
GenBank mRNA AL833251.1 - 25 2640
BC144350.1 - 6 751 9052870
BX647358.1 - 2 4126

Gene Symbol
NPIPB6
Entrez Gene ID
728741
Gene Name
nuclear pore complex interacting protein family member B6
Gene Aliases
NPIPB
Chromosome Location
Chr. 16: 28342517 - 28364971 on Build GRCh38
UniGene ID
Hs.726467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005255742.4 XP_005255799.1 8 1377

Gene Symbol
NPIPB8
Entrez Gene ID
728734
Gene Name
nuclear pore complex interacting protein family member B8
Gene Aliases
Chromosome Location
Chr. 16: 28638065 - 28658693 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017023627.1 XP_016879116.1 8 905

Gene Symbol
NPIPB9
Entrez Gene ID
100507607
Gene Name
nuclear pore complex interacting protein family member B9
Gene Aliases
Chromosome Location
Chr. 16: 28735500 - 28772911 on Build GRCh38
UniGene ID
Hs.710214
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017022828.1 XP_016878317.1 9 1468

View Full Product Details