Gene Transcripts

Gene Symbol
SPANXN5
Entrez Gene ID
494197
Gene Name
SPANX family member N5
Gene Aliases
CT11.10, SPANX-N5
Chromosome Location
Chr. X: 52796146 - 52797348 on Build GRCh38
UniGene ID
Hs.535218
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009616.3 NP_001009616.1 2 157

View Full Product Details