Gene Transcripts

Gene Symbol
ATAD3C
Entrez Gene ID
219293
Gene Name
ATPase family, AAA domain containing 3C
Gene Aliases
Chromosome Location
Chr. 1: 1449689 - 1470158 on Build GRCh38
UniGene ID
Hs.724767
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039211.2 NP_001034300.2 1, 2 1074
GenBank mRNA AK091918.1 - 1, 2 1075

View Full Product Details