Gene Transcripts

Gene Symbol
C15orf62
Entrez Gene ID
643338
Gene Name
chromosome 15 open reading frame 62
Gene Aliases
Chromosome Location
Chr. 15: 40769961 - 40772449 on Build GRCh38
UniGene ID
Hs.631715
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130448.2 NP_001123920.1 1 1645
GenBank mRNA BM706032.1 - 1 314

View Full Product Details