Gene Transcripts
- Gene Symbol
- GOLGA3
- Entrez Gene ID
- 2802
- Gene Name
- golgin A3
- Gene Aliases
- GCP170, MEA-2
- Chromosome Location
- Chr. 12: 132768909 - 132829131 on Build GRCh38
- UniGene ID
- Hs.507333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172557.1 |
NP_001166028.1
|
13, 14 |
3038 |
|
NM_005895.3 |
NP_005886.2
|
13, 14 |
3376 |
|
XM_005266164.4 |
XP_005266221.1
|
13, 14 |
2983 |
|
XM_005266167.4 |
XP_005266224.1
|
10, 11 |
2408 |
|
XM_006719736.3 |
XP_006719799.1
|
13, 14 |
3103 |
|
XM_006719737.3 |
XP_006719800.1
|
13, 14 |
3113 |
|
XM_011534793.2 |
XP_011533095.1
|
12, 13 |
2830 |
|
XM_011534794.2 |
XP_011533096.1
|
5, 6 |
981 |
|
GenBank mRNA |
AB027133.1 |
- |
13, 14 |
3098 |
|
AF485338.1 |
- |
12, 13 |
2820 |
|
BC142658.1 |
- |
13, 14 |
3038 |
40148073 |
BC146675.1 |
- |
13, 14 |
3038 |
40148852 |
D63997.1 |
- |
13, 14 |
3086 |
|
L06148.1 |
- |
3, 4 |
510 |
|
View Full Product Details