Gene Transcripts
- Gene Symbol
- OR2A20P
- Entrez Gene ID
- 401428
- Gene Name
- olfactory receptor family 2 subfamily A member 20 pseudogene
- Gene Aliases
- OR2A20
- Chromosome Location
- Chr. 7: 144250674 - 144251603 on Build GRCh38
- UniGene ID
- Hs.490448
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002158.1 |
|
1 |
818 |
|
GenBank mRNA |
AK172840.1 |
- |
1 |
2172 |
|
BC016940.1 |
- |
3 |
1246 |
|
- Gene Symbol
- OR2A9P
- Entrez Gene ID
- 441295
- Gene Name
- olfactory receptor family 2 subfamily A member 9 pseudogene
- Gene Aliases
- FKSG35, HSDJ0798C17, OR2A19, OR2A22P, OR2A9
- Chromosome Location
- Chr. 7: 144299521 - 144300505 on Build GRCh38
- UniGene ID
- Hs.591830
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002157.1 |
|
1 |
670 |
|
View Full Product Details