Gene Transcripts

Gene Symbol
Bmp7
Entrez Gene ID
12162
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP1
Chromosome Location
Chr. 2: 172868012 - 172940321 on Build GRCm38
UniGene ID
Mm.392150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007557.3 NP_031583.2 3 921
GenBank mRNA BC010771.1 - 3 893
CA316109.1 - 1 56
CJ164635.1 - 2 94
X56906.1 - 3 772

View Full Product Details