Gene Transcripts

Gene Symbol
Cacybp
Entrez Gene ID
12301
Gene Name
calcyclin binding protein
Gene Aliases
SIP
Chromosome Location
Chr. 1: 160202367 - 160212892 on Build GRCm38
UniGene ID
Mm.10702
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009786.2 NP_033916.1 6 854
GenBank mRNA AK013924.1 - 6 785
AK089043.1 - 6 708
AK145013.1 - 6 821
AK215280.1 - 4 318
BC025948.1 - 6 739 5150537
U97327.1 - 6 684

View Full Product Details