Gene Transcripts

Gene Symbol
Cast
Entrez Gene ID
12380
Gene Name
calpastatin
Gene Aliases
Chromosome Location
Chr. 13: 74692366 - 74808874 on Build GRCm38
UniGene ID
Mm.29163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301153.1 NP_001288082.1 11 987
NM_001301155.1 NP_001288084.1 9 718
NM_001301156.1 NP_001288085.1 8 661
NM_001301157.1 NP_001288086.1 8 661
NM_009817.4 NP_033947.1 10 747
GenBank mRNA AB026997.1 - 11 986
AK028841.1 - 10 727
AK028843.1 - 10 709
AK029293.1 - 9 634
AK036576.1 - 8 532
AK083310.1 - 8 564
AK083395.1 - 8 553
AK089108.1 - 8 559
BC010603.1 - 8 515 3710078
CN529927.1 - 9 567
X62519.1 - 7 431

View Full Product Details