Gene Transcripts
- Gene Symbol
- Cbx5
- Entrez Gene ID
- 12419
- Gene Name
- chromobox 5
- Gene Aliases
- 2610029O15Rik, C75991, HP1, Hp1a, Hp1alpha
- Chromosome Location
- Chr. 15: 103191546 - 103239904 on Build GRCm38
- UniGene ID
- Mm.262059
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001076789.1 |
NP_001070257.1
|
2, 3 |
276 |
|
NM_001110216.1 |
NP_001103686.1
|
3, 4 |
377 |
|
NM_007626.3 |
NP_031652.1
|
2, 3 |
277 |
|
GenBank mRNA |
AK008792.1 |
- |
2, 3 |
312 |
|
AK018349.1 |
- |
2, 3 |
223 |
|
AK030366.1 |
- |
2, 3 |
277 |
|
AK030442.1 |
- |
3, 4 |
330 |
|
AK032975.1 |
- |
2, 3 |
233 |
|
BC004707.1 |
- |
2, 3 |
276 |
3499489 |
X99641.1 |
- |
2, 3 |
209 |
|
View Full Product Details