Gene Transcripts

Gene Symbol
Atn1
Entrez Gene ID
13498
Gene Name
atrophin 1
Gene Aliases
Atr1, Drpla, atrophin-1
Chromosome Location
Chr. 6: 124742544 - 124756487 on Build GRCm38
UniGene ID
Mm.333380
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007881.4 NP_031907.2 5 1592
GenBank mRNA BC050920.1 - 5 1592
BC053051.1 - 5 1420 5706586
D87744.1 - 4 1302

View Full Product Details