Gene Transcripts

Gene Symbol
Nkx2-6
Entrez Gene ID
18092
Gene Name
NK2 homeobox 6
Gene Aliases
AV044928, Nkx-2.6, Nkx2.6, Tix, tinman
Chromosome Location
Chr. 14: 69171802 - 69175540 on Build GRCm38
UniGene ID
Mm.390691
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010920.2 NP_035050.2 2 967
GenBank mRNA AF030113.1 - 2 780
AK006401.1 - 2 823
AK007304.1 - 2 750
BC100425.1 - 2 785

View Full Product Details