Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162415.1 | NP_001155887.1 | 10 | 956 | |
NM_001162416.1 | NP_001155888.1 | 8 | 749 | ||
NM_008825.4 | NP_032851.2 | 9 | 850 | ||
GenBank mRNA | AK029659.1 | - | 10 | 957 | |
AK146744.1 | - | 9 | 844 | ||
BC018418.1 | - | 9 | 807 | 3979400 | |
BC051014.1 | - | 8 | 763 | 6493944 | |
EU616666.1 | - | 7 | 623 | ||
EU616667.1 | - | 8 | 748 | ||
EU616668.1 | - | 8 | 748 | ||
EU616669.1 | - | 8 | 748 | ||
EU616670.1 | - | 7 | 614 |