Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008969.4 | NP_032995.1 | 9 | 1288 | |
GenBank mRNA | AK046457.1 | - | 9 | 1215 | |
AK134163.1 | - | 9 | 1174 | ||
AK138432.1 | - | 9 | 1227 | ||
AK142741.1 | - | 9 | 1215 | ||
AK159907.1 | - | 9 | 1289 | ||
AK160886.1 | - | 9 | 1207 | ||
AK163609.1 | - | 9 | 1215 | ||
AK170414.1 | - | 9 | 1183 | ||
AK170664.1 | - | 9 | 1215 | ||
BC005573.1 | - | 9 | 1205 | 3968101 | |
BC023322.1 | - | 9 | 1197 | 3968136 | |
CT010314.1 | - | 8 | 1151 | ||
M34141.1 | - | 9 | 1187 |