Gene Transcripts

Gene Symbol
Ptprg
Entrez Gene ID
19270
Gene Name
protein tyrosine phosphatase, receptor type, G
Gene Aliases
5430405N12Rik, AW046354, AW549872, RPTPgamma
Chromosome Location
Chr. 14: 11553553 - 12242039 on Build GRCm38
UniGene ID
Mm.431266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008981.3 NP_033007.2 10 1946
GenBank mRNA BC079595.1 - 10 1822
L09562.1 - 10 1487

View Full Product Details