Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252089.1 | NP_001239018.1 | 2 | 341 | |
NM_009033.2 | NP_033059.2 | 2 | 387 | ||
GenBank mRNA | AK041927.1 | - | 2 | 278 | |
AK088213.1 | - | 2 | 274 | ||
AK149658.1 | - | 2 | 360 | ||
AK149764.1 | - | 2 | 360 | ||
AK218950.1 | - | 1 | 137 | ||
BC011441.1 | - | 2 | 292 | 3153831 | |
BC089350.1 | - | 2 | 301 | 30359767 | |
BY262810.1 | - | 2 | 341 |