Gene Transcripts

Gene Symbol
Slc50a1
Entrez Gene ID
19729
Gene Name
solute carrier family 50 (sugar transporter), member 1
Gene Aliases
MmSWEET1, Rag1ap1, Rga
Chromosome Location
Chr. 3: 89268246 - 89270570 on Build GRCm38
UniGene ID
Mm.17958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009057.3 NP_033083.2 5 563
GenBank mRNA AK014305.1 - 5 516
AK045924.1 - 3 876
AK079716.1 - 4 628
AK158862.1 - 4 588
AK170162.1 - 5 514
BC014292.1 - 5 547 4190124
X96618.1 - 5 554

View Full Product Details