Gene Transcripts

Gene Symbol
Slc10a1
Entrez Gene ID
20493
Gene Name
solute carrier family 10 (sodium/bile acid cotransporter family), member 1
Gene Aliases
Ntcp
Chromosome Location
Chr. 12: 80953185 - 80968702 on Build GRCm38
UniGene ID
Mm.486361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177561.1 NP_001171032.1 1 237
NM_011387.2 NP_035517.1 1 237
GenBank mRNA AB003303.1 - 1 162
BC021154.1 - 1 199 4162098
BC094023.1 - 2 310 4160598
U95131.1 - 1 142
U95132.1 - 1 142

View Full Product Details