Gene Transcripts

Gene Symbol
Slfn4
Entrez Gene ID
20558
Gene Name
schlafen 4
Gene Aliases
AI450778
Chromosome Location
Chr. 11: 83175171 - 83190216 on Build GRCm38
UniGene ID
Mm.38192
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK137559.1 - 7 1481
AK137689.1 - 6 1402
BC044865.1 - 7 1490 4459564
BC150770.1 - 4 1097 9087681

View Full Product Details