Gene Transcripts

Gene Symbol
Stk10
Entrez Gene ID
20868
Gene Name
serine/threonine kinase 10
Gene Aliases
Gek1, Lok, mKIAA4026
Chromosome Location
Chr. 11: 32533266 - 32624595 on Build GRCm38
UniGene ID
Mm.8235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009288.2 NP_033314.2 14 2300
GenBank mRNA AK148118.1 - 14 2243
BC069840.1 - 14 2373
BC096624.1 - 14 2385

View Full Product Details