Gene Transcripts

Gene Symbol
Tiam1
Entrez Gene ID
21844
Gene Name
T cell lymphoma invasion and metastasis 1
Gene Aliases
AI847750, D16Ium10, D16Ium10e
Chromosome Location
Chr. 16: 89787111 - 90145483 on Build GRCm38
UniGene ID
Mm.310902
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145886.1 NP_001139358.1 25 4432
NM_001145887.1 NP_001139359.1 10 1365
NM_009384.3 NP_033410.2 25 4467
GenBank mRNA AK034803.1 - 3 112
AK136856.1 - 10 1365
AK147392.1 - 25 4433
AK161477.1 - 16 1774
AK162945.1 - 12 1421
BC040748.1 - 3 187
U05245.1 - 25 4470

View Full Product Details