Gene Transcripts

Gene Symbol
Tsx
Entrez Gene ID
22127
Gene Name
testis specific X-linked gene
Gene Aliases
AV207184
Chromosome Location
Chr. X: 103409101 - 103424588 on Build GRCm38
UniGene ID
Mm.358644
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009440.2 NP_033466.1 5 389
GenBank mRNA AK018925.1 - 1 30
BC048493.1 - 5 389 6705048
X99796.1 - 5 374

View Full Product Details