Gene Transcripts

Gene Symbol
Usp9x
Entrez Gene ID
22284
Gene Name
ubiquitin specific peptidase 9, X chromosome
Gene Aliases
5730589N07Rik, AA407302, AA407699, AL022658, AL022749, Dffrx, FAF-X, Fafl
Chromosome Location
Chr. X: 13070997 - 13173328 on Build GRCm38
UniGene ID
Mm.242646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009481.2 NP_033507.2 35 5783
GenBank mRNA CF733628.1 - 5 649
DQ086491.1 - 35 5744
U67874.1 - 35 5744

View Full Product Details