Gene Transcripts
- Gene Symbol
- Vpreb2
- Entrez Gene ID
- 22363
- Gene Name
- pre-B lymphocyte gene 2
- Gene Aliases
- CD179b, VpreB1, Vpreb-2
- Chromosome Location
- Chr. 16: 17979452 - 17981274 on Build GRCm38
- UniGene ID
- Mm.377118
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016983.1 |
NP_058679.1
|
2 |
313 |
|
- Gene Symbol
- Vpreb1
- Entrez Gene ID
- 22362
- Gene Name
- pre-B lymphocyte gene 1
- Gene Aliases
- CD179a, Vpreb-1
- Chromosome Location
- Chr. 16: 16868401 - 16869255 on Build GRCm38
- UniGene ID
- Mm.57574
- Species
- Mus musculus
View Full Product Details