Gene Transcripts

Gene Symbol
Wnt8b
Entrez Gene ID
22423
Gene Name
wingless-type MMTV integration site family, member 8B
Gene Aliases
Chromosome Location
Chr. 19: 44493472 - 44514273 on Build GRCm38
UniGene ID
Mm.88365
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011720.3 NP_035850.2 4 427
GenBank mRNA AF130349.1 - 4 429
AK050992.1 - 4 428
BC119544.2 - 4 359 40056929

View Full Product Details