Gene Transcripts
- Gene Symbol
- Map3k2
- Entrez Gene ID
- 26405
- Gene Name
- mitogen-activated protein kinase kinase kinase 2
- Gene Aliases
- 9630061B06Rik, AI585793, Mekk2, Mekk2b
- Chromosome Location
- Chr. 18: 32163089 - 32236751 on Build GRCm38
- UniGene ID
- Mm.211762
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011946.3 |
NP_036076.2
|
10, 11 |
1271 |
|
GenBank mRNA |
U43186.1 |
- |
10, 11 |
1169 |
|
View Full Product Details