Gene Transcripts

Gene Symbol
Phlda3
Entrez Gene ID
27280
Gene Name
pleckstrin homology-like domain, family A, member 3
Gene Aliases
Tih1
Chromosome Location
Chr. 1: 135766085 - 135769134 on Build GRCm38
UniGene ID
Mm.34346
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013750.2 NP_038778.1 2 1195
GenBank mRNA AK003767.1 - 2 1161
BC023376.1 - 2 1104
BC023408.1 - 2 1104 5252055

View Full Product Details