Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021299.1 | NP_067274.1 | 5 | 663 | |
GenBank mRNA | AB020203.1 | - | 5 | 566 | |
AK004864.2 | - | 5 | 678 | ||
AK005194.2 | - | 5 | 717 | ||
AK008681.1 | - | 5 | 680 | ||
AK031631.1 | - | 5 | 661 | ||
AK053644.1 | - | 5 | 723 | ||
AK089027.1 | - | 5 | 714 | ||
AK147060.1 | - | 5 | 723 | ||
AK149996.1 | - | 5 | 720 | ||
AK207955.1 | - | 46 | |||
BC016432.1 | - | 5 | 665 | 4217820 | |
BC019174.1 | - | 5 | 716 | 5007578 | |
BC024871.1 | - | 5 | 663 | 3154594 | |
BC058191.1 | - | 5 | 693 | 5340253 |