Gene Transcripts

Gene Symbol
Capn12
Entrez Gene ID
60594
Gene Name
calpain 12
Gene Aliases
CANP 12
Chromosome Location
Chr. 7: 28881446 - 28893585 on Build GRCm38
UniGene ID
Mm.476990
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110807.1 NP_001104277.1 8 1008
GenBank mRNA AJ289243.1 - 8 1016
BC028751.1 - 8 1007 1314413

View Full Product Details