Gene Transcripts

Gene Symbol
Oxct2b
Entrez Gene ID
353371
Gene Name
3-oxoacid CoA transferase 2B
Gene Aliases
Scot-t2
Chromosome Location
Chr. 4: 123116248 - 123118000 on Build GRCm38
UniGene ID
Mm.189660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181859.3 NP_862907.2 1 1056
GenBank mRNA AB049996.1 - 1 1046
BC113758.1 - 1 1039 40047052

Gene Symbol
Oxct2a
Entrez Gene ID
64059
Gene Name
3-oxoacid CoA transferase 2A
Gene Aliases
Oxct, Oxct2, Scot, Scot-t1
Chromosome Location
Chr. 4: 123321875 - 123323679 on Build GRCm38
UniGene ID
Mm.270287
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022033.4 NP_071316.1 1 1108
GenBank mRNA AB022180.2 - 1 1046
AB045847.1 - 1 127
AK077075.1 - 1 1061
BC137887.1 - 1 1108 8860906
BC137888.1 - 1 1108 8860907

View Full Product Details