Gene Transcripts

Gene Symbol
Alkbh7
Entrez Gene ID
66400
Gene Name
alkB, alkylation repair homolog 7 (E. coli)
Gene Aliases
2310045B01Rik, 2510008E23Rik, Abh7, Spata11
Chromosome Location
Chr. 17: 56997339 - 56999340 on Build GRCm38
UniGene ID
Mm.196150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025538.3 NP_079814.1 3 517
NM_027372.1 NP_081648.1 2 343
GenBank mRNA AK007380.1 - 3 495
AK009812.1 - 3 504
AK010930.1 - 2 344
BC029677.1 - 3 484 5364485

View Full Product Details