Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025597.2 | NP_079873.1 | 3 | 283 | |
GenBank mRNA | AK007930.1 | - | 3 | 282 | |
AK009008.1 | - | 3 | 245 | ||
AK012320.1 | - | 3 | 284 | ||
AK013608.1 | - | 3 | 269 | ||
AK138123.1 | - | 3 | 281 | ||
BC028669.1 | - | 3 | 439 | ||
BC036989.1 | - | 3 | 280 | 4989233 |