Gene Transcripts
- Gene Symbol
- Ubr7
- Entrez Gene ID
- 66622
- Gene Name
- ubiquitin protein ligase E3 component n-recognin 7 (putative)
- Gene Aliases
- 5730410I19Rik, AA589405, AW557761
- Chromosome Location
- Chr. 12: 102757975 - 102777701 on Build GRCm38
- UniGene ID
- Mm.34261
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025666.2 |
NP_079942.1
|
10, 11 |
1243 |
|
GenBank mRNA |
AK088208.1 |
- |
10, 11 |
1245 |
|
AK146308.1 |
- |
10, 11 |
1245 |
|
AK149989.1 |
- |
10, 11 |
1198 |
|
AK155611.1 |
- |
10, 11 |
1114 |
|
BC051678.1 |
- |
7, 8 |
771 |
|
BC058535.1 |
- |
9, 10 |
927 |
|
BC079531.1 |
- |
9, 10 |
940 |
|
BC094323.1 |
- |
10, 11 |
1250 |
|
BC094423.1 |
- |
10, 11 |
1201 |
6812389 |
View Full Product Details