Gene Transcripts
- Gene Symbol
- 9030624G23Rik
- Entrez Gene ID
- 66808
- Gene Name
- RIKEN cDNA 9030624G23 gene
- Gene Aliases
- Chromosome Location
- Chr. 12: 24043202 - 24097269 on Build GRCm38
- UniGene ID
- Mm.379124
- Species
- Mus musculus
- Gene Symbol
- Asap2
- Entrez Gene ID
- 211914
- Gene Name
- ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
- Gene Aliases
- 6530401G17Rik, Ddef2, Gm1523, Gm592, PAG3, PAP
- Chromosome Location
- Chr. 12: 21111713 - 21270171 on Build GRCm38
- UniGene ID
- Mm.358946
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018309.1 |
- |
|
649 |
|
- Gene Symbol
- Gm9222
- Entrez Gene ID
- 668525
- Gene Name
- predicted gene 9222
- Gene Aliases
- BDR-1, CA748277, EG668383, EG668525
- Chromosome Location
- Chr. 12: 17881914 - 17939655 on Build GRCm38
- UniGene ID
- Mm.278091
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC031503.1 |
- |
|
817 |
|
View Full Product Details