Gene Transcripts

Gene Symbol
Gatad1
Entrez Gene ID
67210
Gene Name
GATA zinc finger domain containing 1
Gene Aliases
2310031E19Rik, 2810047M21Rik, 8430439A17Rik, 9130430G15Rik, B330017N08Rik, ENSMUSG00000073241, Odag
Chromosome Location
Chr. 5: 3639961 - 3647936 on Build GRCm38
UniGene ID
Mm.178424
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026033.2 NP_080309.2 4 904
GenBank mRNA AB047921.1 - 4 885
AK078416.1 - 4 849
BC019449.1 - 4 774 4024452

View Full Product Details