Gene Transcripts

Gene Symbol
MARK1
Entrez Gene ID
4139
Gene Name
microtubule affinity regulating kinase 1
Gene Aliases
MARK, Par-1c, Par1c
Chromosome Location
Chr. 1: 220528183 - 220664457 on Build GRCh38
UniGene ID
Hs.497806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286124.1 NP_001273053.1 13 2036
NM_001286126.1 NP_001273055.1 13 2036
NM_001286128.1 NP_001273057.1 12 1970
NM_018650.4 NP_061120.3 13 2036
XM_005273134.4 XP_005273191.1 13 1903
XM_006711326.3 XP_006711389.1 13 1297
XM_011509561.2 XP_011507863.1 13 2032
XM_011509562.2 XP_011507864.1 13 1649
XM_017001305.1 XP_016856794.1 14 1493
XM_017001306.1 XP_016856795.1 13 1533
XM_017001307.1 XP_016856796.1 14 1933
GenBank mRNA AB040910.1 - 12 1927
AF154845.1 - 13 1662
AK027493.1 - 9 993
AK295506.1 - 13 1534
AK314614.1 - 13 1696
BC113869.1 - 13 1572 40036731
BC114478.1 - 13 1572 40036730
KJ534879.1 - 13 1396
KJ534880.1 - 7 796
KJ534881.1 - 13 1396
KJ535089.1 - 13 1395

View Full Product Details