Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026329.2 | NP_080605.1 | 2 | 150 | |
GenBank mRNA | AK007892.1 | - | 2 | 171 | |
AK010687.1 | - | 2 | 119 | ||
AK011627.1 | - | 2 | 151 | ||
AK013134.1 | - | 2 | 121 | ||
AK017473.1 | - | 2 | 129 | ||
AK020719.1 | - | 2 | 119 | ||
BC005580.1 | - | 2 | 169 | 3968561 | |
BC055278.1 | - | 2 | 100 | 5699685 |