Gene Transcripts

Gene Symbol
MFNG
Entrez Gene ID
4242
Gene Name
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
Chromosome Location
Chr. 22: 37469063 - 37486440 on Build GRCh38
UniGene ID
Hs.517603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166343.1 NP_001159815.1 4 775
NM_002405.3 NP_002396.2 5 870
NR_029413.1 4 821
GenBank mRNA AK292123.1 - 5 811
AK297149.1 - 4 769
AK297833.1 - 4 754
AK316555.1 - 4 762
BC094814.1 - 5 831 5930073
BM457814.1 - 5 827
CR456518.1 - 5 663
U94352.1 - 5 777

View Full Product Details