Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027120.2 | NP_081396.1 | 7 | 596 | |
GenBank mRNA | AK009352.1 | - | 7 | 612 | |
AK137483.1 | - | 7 | 598 | ||
BC119244.1 | - | 7 | 558 | 8733993 | |
BC119246.1 | - | 7 | 558 | 8733995 |