Gene Transcripts

Gene Symbol
Ndufaf1
Entrez Gene ID
69702
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1
Gene Aliases
2410001M24Rik, CGI-65, CIA30
Chromosome Location
Chr. 2: 119655446 - 119662885 on Build GRCm38
UniGene ID
Mm.5390
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027175.3 NP_081451.3 3 959
GenBank mRNA AK010328.1 - 2 789
AK050343.1 - 3 960
BC018422.1 - 3 789 3593650

View Full Product Details