Gene Transcripts

Gene Symbol
MLN
Entrez Gene ID
4295
Gene Name
motilin
Gene Aliases
Chromosome Location
Chr. 6: 33794672 - 33804016 on Build GRCh38
UniGene ID
Hs.2813
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040109.1 NP_001035198.1 2, 3 196
NM_001184698.1 NP_001171627.1 2, 3 196
NM_002418.2 NP_002409.1 2, 3 196
GenBank mRNA BC069675.1 - 2, 3 191
BC112314.1 - 2, 3 160 8327782
BC143981.1 - 2, 3 160 9052499
Y00695.1 - 2, 3 180

View Full Product Details