Gene Transcripts

Gene Symbol
Ankib1
Entrez Gene ID
70797
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
2310061P20Rik, 4631416I11Rik, AW494740, C80642, mKIAA1386
Chromosome Location
Chr. 5: 3689999 - 3803124 on Build GRCm38
UniGene ID
Mm.130148
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003909.4 NP_001003909.2 13 2419
NM_001289527.1 NP_001276456.1 13 2419
NM_001289528.1 NP_001276457.1 12 2301
NM_001289529.1 NP_001276458.1 13 2028
GenBank mRNA AK129343.1 - 13 2220
BC020475.1 - 13 1953
BC079620.1 - 2 109
BC089031.1 - 13 2007
BC145510.1 - 12 2052
BC145511.1 - 13 2170 9054074
BC150792.1 - 13 2170 9087703

View Full Product Details